Search
Search

PYR4| Gene Symbol: PYR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341566.1 | NP_001327985.1 | 3 - 4 | 355 | 65 | |
| NM_118422.4 | NP_194024.1 | 3 - 4 | 355 | 65 | ||
| GenBank mRNA | AF000146.1 | - | 3 - 4 | 265 | 65 | |
Set Membership: |
|