Search
Search

ENDO4| Gene Symbol: ENDO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341501.1 | NP_001328108.1 | 3 - 4 | 711 | 78 | |
| NM_001341502.1 | NP_001328109.1 | 3 - 4 | 683 | 78 | ||
| NM_001341503.1 | NP_001328107.1 | 5 - 6 | 493 | 78 | ||
| NM_001341504.1 | NP_001328110.1 | 3 - 4 | 663 | 78 | ||
| NM_001341505.1 | NP_001328111.1 | 4 - 5 | 585 | 78 | ||
| NM_148368.2 | NP_680734.1 | 4 - 5 | 633 | 78 | ||
Set Membership: |
|