Search
Search

INT2| Gene Symbol: INT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102761.4 | NP_174313.1 | 3 - 4 | 688 | 80 | |
| GenBank mRNA | AJ973176.1 | - | 3 - 4 | 546 | 80 | |
| AY074333.1 | - | 3 - 4 | 578 | 80 | ||
| AY123031.1 | - | 3 - 4 | 546 | 80 | ||
Set Membership: |
|