Search
Search

CCC1| Gene Symbol: CCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102781.3 | NP_174333.2 | 3 - 4 | 582 | 113 | |
| NM_179400.1 | NP_849731.1 | 3 - 4 | 551 | 113 | ||
| NM_179401.2 | NP_849732.1 | 3 - 4 | 582 | 113 | ||
| GenBank mRNA | AK226602.1 | - | 3 - 4 | 537 | 113 | |
| AM113986.1 | - | 3 - 4 | 552 | 113 | ||
Set Membership: |
|