Search
Search

AT1G30580| Gene Symbol: AT1G30580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102795.3 | NP_174346.1 | 11 - 12 | 1265 | 141 | |
| GenBank mRNA | BT025965.1 | - | 11 - 12 | 1084 | 141 | |
Set Membership: |
|