Search
Search

GlcNAc1pUT1| Gene Symbol: GlcNAc1pUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332942.1 | NP_001319120.1 | 5 - 6 | 838 | 103 | |
| NM_102845.5 | NP_564372.3 | 5 - 6 | 838 | 103 | ||
| GenBank mRNA | AY053411.1 | - | 5 - 6 | 644 | 103 | |
Set Membership: |
|