Search
Search

ABCG14| Gene Symbol: ABCG14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102911.3 | NP_564383.1 | 3 - 4 | 1344 | 67 | |
| GenBank mRNA | AK117530.1 | - | 3 - 4 | 1253 | 67 | |
| AY088793.1 | - | 3 - 4 | 1243 | 67 | ||
Set Membership: |
|