Search
Search

ATS1| Gene Symbol: ATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102953.4 | NP_174499.1 | 2 - 3 | 585 | 78 | |
| NM_179407.3 | NP_849738.1 | 2 - 3 | 585 | 78 | ||
| GenBank mRNA | AY093169.1 | - | 2 - 3 | 470 | 78 | |
| BT008758.1 | - | 2 - 3 | 412 | 78 | ||
Set Membership: |
|