Search
Search

sks9| Gene Symbol: sks9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342497.1 | NP_001329210.1 | 5 - 6 | 1428 | 70 | |
| NM_120004.5 | NP_195555.2 | 5 - 6 | 1586 | 70 | ||
| GenBank mRNA | AY072215.1 | - | 5 - 6 | 1428 | 70 | |
| AY096593.1 | - | 5 - 6 | 1362 | 70 | ||
Set Membership: |
|