Search
Search

AT4G38550| Gene Symbol: AT4G38550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342501.1 | NP_001328678.1 | 1 - 2 | 50 | 83 | |
| NM_001342502.1 | NP_001328679.1 | 1 - 2 | 50 | 83 | ||
| NM_120016.6 | NP_195567.2 | 2 - 3 | 208 | 83 | ||
| GenBank mRNA | AF360282.1 | - | 2 - 3 | 208 | 83 | |
| BT000695.1 | - | 2 - 3 | 141 | 83 | ||
| BT001966.1 | - | 1 - 2 | 50 | 83 | ||
Set Membership: |
|