Search
Search

LIP1| Gene Symbol: LIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127084.3 | NP_179126.2 | 3 - 4 | 578 | 111 | |
| GenBank mRNA | AF530477.1 | - | 3 - 4 | 419 | 111 | |
| AK229578.1 | - | 3 - 4 | 477 | 111 | ||
| BT025547.1 | - | 3 - 4 | 392 | 111 | ||
Set Membership: |
|