Search
Search

PHT3;3| Gene Symbol: PHT3;3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127282.3 | NP_179319.1 | 5 - 6 | 1239 | 99 | |
| GenBank mRNA | AK227300.1 | - | 5 - 6 | 1237 | 99 | |
| BT010199.1 | - | 5 - 6 | 880 | 99 | ||
Set Membership: |
|