Search
Search

AT2G17975| Gene Symbol: AT2G17975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335574.1 | NP_001323512.1 | 5 - 6 | 701 | 62 | |
| NM_127353.3 | NP_179388.1 | 5 - 6 | 826 | 62 | ||
| GenBank mRNA | BT015337.1 | - | 5 - 6 | 710 | 62 | |
| BT015844.1 | - | 5 - 6 | 675 | 62 | ||
Set Membership: |
|