Search
Search

ATCS| Gene Symbol: ATCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337082.1 | NP_001324514.1 | 15 - 16 | 1217 | 110 | |
| NM_001337083.1 | NP_001324515.1 | 15 - 16 | 1125 | 110 | ||
| NM_129998.3 | NP_566016.1 | 15 - 16 | 1166 | 110 | ||
| NM_180084.2 | NP_850415.1 | 15 - 16 | 1169 | 110 | ||
| GenBank mRNA | AF387018.1 | - | 15 - 16 | 1168 | 110 | |
| BT006613.1 | - | 14 - 15 | 1034 | 110 | ||
Set Membership: |
|