Search
Search

POL| Gene Symbol: POL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337223.1 | NP_001325392.1 | 2 - 3 | 2829 | 139 | |
| NM_001337224.1 | NP_001325391.1 | 2 - 3 | 2770 | 139 | ||
| NM_180132.3 | NP_850463.1 | 2 - 3 | 2835 | 139 | ||
| NM_180133.2 | NP_850464.1 | 1 - 2 | 2935 | 139 | ||
Set Membership: |
|