Search
Search

AT2G47310| Gene Symbol: AT2G47310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180141.2 | NP_850472.1 | 8 - 9 | 1019 | 112 | |
| GenBank mRNA | AK229352.1 | - | 8 - 9 | 920 | 112 | |
| BT015132.1 | - | 8 - 9 | 884 | 112 | ||
| BT015799.1 | - | 8 - 9 | 817 | 112 | ||
Set Membership: |
|