Search
Search

AT2G47500| Gene Symbol: AT2G47500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337265.1 | NP_001325363.1 | 7 - 8 | 1224 | 131 | |
| NM_180144.4 | NP_850475.2 | 7 - 8 | 1266 | 131 | ||
Set Membership: |
|