Search
Search

SWI3A| Gene Symbol: SWI3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337268.1 | NP_001324817.1 | 3 - 4 | 1125 | 87 | |
| NM_180145.3 | NP_850476.1 | 3 - 4 | 1117 | 87 | ||
| GenBank mRNA | AY062794.1 | - | 3 - 4 | 1096 | 87 | |
| AY081570.1 | - | 3 - 4 | 1030 | 87 | ||
Set Membership: |
|