Search
Search

RPH1| Gene Symbol: RPH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202847.1 | NP_001189776.1 | 2 - 3 | 441 | 65 | |
| NM_130374.4 | NP_566122.1 | 2 - 3 | 543 | 65 | ||
| NM_180153.2 | NP_850484.1 | 2 - 3 | 414 | 65 | ||
| GenBank mRNA | AY045783.1 | - | 2 - 3 | 408 | 65 | |
| AY079358.1 | - | 2 - 3 | 395 | 65 | ||
Set Membership: |
|