Search
Search

RR22| Gene Symbol: RR22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084633.1 | NP_001078102.1 | 2 - 3 | 347 | 114 | |
| NM_111299.3 | NP_187078.1 | 2 - 3 | 479 | 114 | ||
| NM_180180.3 | NP_850511.1 | 1 - 2 | 662 | 114 | ||
| GenBank mRNA | AY085638.1 | - | 2 - 3 | 464 | 114 | |
Set Membership: |
|