Search
Search

IDH-VI| Gene Symbol: IDH-VI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180218.2 | NP_850549.1 | 5 - 6 | 1110 | 80 | |
| GenBank mRNA | AF324664.2 | - | 5 - 6 | 915 | 80 | |
| AF327427.1 | - | 5 - 6 | 915 | 80 | ||
| AF339723.1 | - | 5 - 6 | 856 | 80 | ||
| AK176269.1 | - | 5 - 6 | 915 | 80 | ||
| AK228113.1 | - | 5 - 6 | 915 | 80 | ||
Set Membership: |
|