Search
Search

AT3G11960| Gene Symbol: AT3G11960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337966.1 | NP_001327782.1 | 15 - 16 | 4345 | 72 | |
| NM_001337967.1 | NP_001327783.1 | 15 - 16 | 4259 | 72 | ||
| NM_180234.2 | NP_850565.1 | 13 - 14 | 4078 | 72 | ||
Set Membership: |
|