Search
Search

DJ1A| Gene Symbol: DJ1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035621.1 | NP_001030698.1 | 4 - 5 | 671 | 90 | |
| NM_112361.5 | NP_188117.1 | 4 - 5 | 693 | 90 | ||
| NM_180257.1 | NP_850588.1 | 3 - 4 | 584 | 90 | ||
| GenBank mRNA | AF326856.1 | - | 4 - 5 | 618 | 90 | |
| AF349515.1 | - | 4 - 5 | 559 | 90 | ||
| AK317457.1 | - | 4 - 5 | 618 | 90 | ||
| AY039574.1 | - | 4 - 5 | 618 | 90 | ||
| AY129490.1 | - | 4 - 5 | 559 | 90 | ||
Set Membership: |
|