Search
Search

PBP1| Gene Symbol: PBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035633.1 | NP_001030710.1 | 2 - 3 | 299 | 68 | |
| NM_112513.3 | NP_188263.1 | 2 - 3 | 395 | 68 | ||
| NM_180263.2 | NP_850594.1 | 1 - 2 | 286 | 68 | ||
| GenBank mRNA | AF370488.1 | - | 2 - 3 | 264 | 68 | |
| AY064653.1 | - | 1 - 2 | 201 | 68 | ||
| AY065189.1 | - | 2 - 3 | 272 | 68 | ||
| AY128360.1 | - | 1 - 2 | 283 | 68 | ||
| BT000021.1 | - | 1 - 2 | 201 | 68 | ||
| BT002196.1 | - | 1 - 2 | 201 | 68 | ||
Set Membership: |
|