Search
Search

MYB4R1| Gene Symbol: MYB4R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202993.1 | NP_001189922.1 | 6 - 7 | 1313 | 82 | |
| NM_001338329.1 | NP_001327710.1 | 5 - 6 | 1450 | 82 | ||
| NM_112692.3 | NP_566597.1 | 4 - 5 | 1418 | 82 | ||
| NM_180276.2 | NP_850607.1 | 5 - 6 | 1348 | 82 | ||
| GenBank mRNA | AY033827.1 | - | 4 - 5 | 1292 | 82 | |
Set Membership: |
|