Search
Search

JMJD5| Gene Symbol: JMJD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035656.1 | NP_001030733.1 | 2 - 3 | 802 | 78 | |
| NM_001338491.1 | NP_001327066.1 | 2 - 3 | 848 | 78 | ||
| NM_001338492.1 | NP_001327067.1 | 2 - 3 | 820 | 78 | ||
| NM_112972.4 | NP_566667.1 | 2 - 3 | 863 | 78 | ||
| NM_180286.1 | NP_850617.1 | 2 - 3 | 867 | 78 | ||
| GenBank mRNA | AY084468.1 | - | 2 - 3 | 791 | 78 | |
| AY091769.1 | - | 2 - 3 | 867 | 78 | ||
| AY125538.1 | - | 2 - 3 | 727 | 78 | ||
Set Membership: |
|