Search
Search

ARV1| Gene Symbol: ARV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035846.2 | NP_001030923.1 | 4 - 5 | 357 | 88 | |
| NM_001331239.1 | NP_001318899.1 | 4 - 5 | 750 | 88 | ||
| NM_001331240.1 | NP_001321775.1 | 3 - 4 | 841 | 88 | ||
| NM_001331241.1 | NP_001321777.1 | 3 - 4 | 818 | 88 | ||
| NM_001331242.1 | NP_001321776.1 | 4 - 5 | 727 | 88 | ||
| GenBank mRNA | AK228414.1 | - | 4 - 5 | 312 | 88 | |
| AY758070.1 | - | 4 - 5 | 357 | 88 | ||
Set Membership: |
|