Search
Search

FER1| Gene Symbol: FER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120238.4 | NP_195780.1 | 1 - 2 | 768 | 93 | |
| GenBank mRNA | AF326869.1 | - | 1 - 2 | 380 | 93 | |
| AF339691.1 | - | 1 - 2 | 311 | 93 | ||
| AF412065.1 | - | 1 - 2 | 380 | 93 | ||
| AK226433.1 | - | 1 - 2 | 383 | 93 | ||
| X94248.1 | - | 349 | 93 | |||
Set Membership: |
|