Search
Search

SCAR4| Gene Symbol: SCAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342607.1 | NP_001318456.1 | 8 - 9 | 3588 | 75 | |
| NM_001342610.1 | NP_001330637.1 | 7 - 8 | 3679 | 75 | ||
| NM_001342611.1 | NP_001330636.1 | 6 - 7 | 3524 | 75 | ||
| NM_001342612.1 | NP_001330633.1 | 5 - 6 | 3081 | 75 | ||
| NM_001342613.1 | NP_001330635.1 | 2 - 3 | 2590 | 75 | ||
| NM_001342614.1 | NP_001330634.1 | 7 - 8 | 3679 | 75 | ||
| NM_001342615.1 | NP_001330639.1 | 6 - 7 | 3524 | 75 | ||
| NM_001342616.1 | NP_001330638.1 | 7 - 8 | 3429 | 75 | ||
| NM_120251.4 | NP_195793.3 | 7 - 8 | 3470 | 75 | ||
| GenBank mRNA | AY743927.1 | - | 7 - 8 | 3349 | 75 | |
Set Membership: |
|