Search
Search

RAPTOR2| Gene Symbol: RAPTOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342617.1 | NP_001330425.1 | 15 - 16 | 2908 | 68 | |
| NM_001342618.1 | NP_001330424.1 | 15 - 16 | 2908 | 68 | ||
| NM_001342619.1 | NP_001330427.1 | 15 - 16 | 2874 | 68 | ||
| NM_001342620.1 | NP_001330426.1 | 15 - 16 | 2868 | 68 | ||
| NM_120255.3 | NP_195797.3 | 15 - 16 | 2865 | 68 | ||
Set Membership: |
|