Search
Search

RPL| Gene Symbol: RPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120281.3 | NP_195823.1 | 3 - 4 | 1368 | 66 | |
| GenBank mRNA | AY328314.1 | - | 3 - 4 | 1301 | 66 | |
| BT020537.1 | - | 3 - 4 | 1208 | 66 | ||
Set Membership: |
|