Search
Search

OHP| Gene Symbol: OHP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120290.3 | NP_195832.1 | 1 - 2 | 194 | 64 | |
| GenBank mRNA | AF054617.1 | - | 1 - 2 | 153 | 64 | |
| AY062846.1 | - | 1 - 2 | 164 | 64 | ||
| AY081584.1 | - | 1 - 2 | 141 | 64 | ||
Set Membership: |
|