Search
Search

IDH-V| Gene Symbol: IDH-V | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120407.6 | NP_568113.1 | 1 - 2 | 253 | 95 | |
| GenBank mRNA | AF412100.1 | - | 1 - 2 | 244 | 95 | |
| AY099823.1 | - | 1 - 2 | 232 | 95 | ||
| BT008460.1 | - | 1 - 2 | 184 | 95 | ||
Set Membership: |
|