Search
Search

RABC2A| Gene Symbol: RABC2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001342711.1 | NP_001332165.1 | 2 - 3 | 410 | 77 | |
| NM_120433.3 | NP_568121.1 | 1 - 2 | 639 | 77 | ||
| GenBank mRNA | AK119027.1 | - | 1 - 2 | 465 | 77 | |
| BT006101.1 | - | 1 - 2 | 136 | 77 | ||
| D89824.1 | - | 1 - 2 | 166 | 77 | ||
Set Membership: |
|