Search
Search

CTR1| Gene Symbol: CTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120454.4 | NP_195993.1 | 9 - 10 | 2075 | 89 | |
| NM_180429.4 | NP_850760.1 | 9 - 10 | 2075 | 89 | ||
| GenBank mRNA | AK227171.1 | - | 9 - 10 | 2084 | 89 | |
| L08789.1 | - | 9 - 10 | 2003 | 89 | ||
Set Membership: |
|