Search
Search

TFL1| Gene Symbol: TFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120465.3 | NP_196004.1 | 2 - 3 | 349 | 77 | |
| GenBank mRNA | BT024828.1 | - | 2 - 3 | 273 | 77 | |
| U77674.1 | - | 2 - 3 | 288 | 77 | ||
Set Membership: |
|