Search
Search

CPSRP54| Gene Symbol: CPSRP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120476.5 | NP_196014.1 | 4 - 5 | 623 | 72 | |
| GenBank mRNA | AY050999.1 | - | 4 - 5 | 519 | 72 | |
| BT000976.1 | - | 4 - 5 | 455 | 72 | ||
| Z21970.1 | - | 478 | 72 | |||
Set Membership: |
|