Search
Search

AT2G19010| Gene Symbol: AT2G19010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335618.1 | NP_001323983.1 | 4 - 5 | 963 | 101 | |
| NM_127458.2 | NP_179491.2 | 4 - 5 | 901 | 101 | ||
Set Membership: |
|