Search
Search

ckl5| Gene Symbol: ckl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127505.5 | NP_179537.1 | 12 - 13 | 1297 | 117 | |
| GenBank mRNA | AY086262.1 | - | 12 - 13 | 1229 | 117 | |
| AY099837.1 | - | 12 - 13 | 1188 | 117 | ||
| AY943844.1 | - | 12 - 13 | 990 | 117 | ||
| BT006580.1 | - | 12 - 13 | 990 | 117 | ||
Set Membership: |
|