Search
Search

AT2G20400| Gene Symbol: AT2G20400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335680.1 | NP_001323847.1 | 3 - 4 | 1050 | 95 | |
| NM_127599.3 | NP_179630.2 | 3 - 4 | 1095 | 95 | ||
| GenBank mRNA | AK118313.1 | - | 3 - 4 | 949 | 95 | |
| BT006119.1 | - | 2 - 3 | 758 | 95 | ||
Set Membership: |
|