Search
Search

NOP10| Gene Symbol: NOP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036302.1 | NP_001031379.1 | 3 - 4 | 188 | 97 | |
| NM_001335686.1 | NP_001325224.1 | 2 - 3 | 281 | 97 | ||
| NM_127608.4 | NP_565472.1 | 3 - 4 | 226 | 97 | ||
| GenBank mRNA | AY087879.1 | - | 3 - 4 | 185 | 97 | |
Set Membership: |
|