Search
Search

AT5G17410| Gene Symbol: AT5G17410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121747.4 | NP_568346.1 | 7 - 8 | 723 | 98 | |
| NM_180507.2 | NP_850838.1 | 7 - 8 | 685 | 98 | ||
| GenBank mRNA | AF360238.1 | - | 7 - 8 | 682 | 98 | |
| AY040037.1 | - | 7 - 8 | 641 | 98 | ||
Set Membership: |
|