Search
Search

AT1G22700| Gene Symbol: AT1G22700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102117.5 | NP_564178.2 | 1 - 2 | 380 | 86 | |
| NM_180619.2 | NP_850950.1 | 1 - 2 | 206 | 86 | ||
| GenBank mRNA | AF410279.1 | - | 1 - 2 | 212 | 86 | |
| AK317512.1 | - | 1 - 2 | 207 | 86 | ||
| BT001148.1 | - | 1 - 2 | 122 | 86 | ||
Set Membership: |
|