Search
Search

EGY2| Gene Symbol: EGY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203303.1 | NP_001190232.1 | 11 - 12 | 1493 | 127 | |
| NM_120656.3 | NP_196193.1 | 10 - 11 | 1491 | 127 | ||
| NM_203007.3 | NP_974736.1 | 11 - 12 | 1540 | 127 | ||
Set Membership: |
|