Search
Search

NUDX27| Gene Symbol: NUDX27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120717.5 | NP_196252.1 | 3 - 4 | 500 | 90 | |
| GenBank mRNA | AF372941.1 | - | 3 - 4 | 433 | 90 | |
| AY113163.1 | - | 3 - 4 | 372 | 90 | ||
Set Membership: |
|