Search
Search

GUS2| Gene Symbol: GUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120865.5 | NP_196400.2 | 4 - 5 | 845 | 64 | |
| GenBank mRNA | BT015345.1 | - | 4 - 5 | 626 | 64 | |
| BT015800.1 | - | 3 - 4 | 446 | 64 | ||
Set Membership: |
|