Search
Search

NF-YB12| Gene Symbol: NF-YB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_120902.3 | NP_568190.1 | 5 - 6 | 588 | 70 | |
| GenBank mRNA | AF428400.1 | - | 5 - 6 | 473 | 70 | |
| AK227164.1 | - | 5 - 6 | 507 | 70 | ||
| AY087017.1 | - | 5 - 6 | 514 | 70 | ||
| AY125544.1 | - | 4 - 5 | 316 | 70 | ||
Set Membership: |
|