Search
Search

PROPEP6| Gene Symbol: PROPEP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127769.3 | NP_179791.1 | 1 - 2 | 510 | 141 | |
| GenBank mRNA | BT021966.1 | - | 1 - 2 | 295 | 141 | |
| BT024750.1 | - | 1 - 2 | 215 | 141 | ||
Set Membership: |
|