Search
Search

PMDH1| Gene Symbol: PMDH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127843.5 | NP_179863.1 | 7 - 8 | 1135 | 72 | |
| GenBank mRNA | AF428346.1 | - | 7 - 8 | 1047 | 72 | |
| AK227067.1 | - | 7 - 8 | 1045 | 72 | ||
| BT003009.1 | - | 7 - 8 | 904 | 72 | ||
Set Membership: |
|