Search
Search

PAB4| Gene Symbol: PAB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127899.4 | NP_179916.1 | 1 - 2 | 593 | 148 | |
| GenBank mRNA | AY050859.1 | - | 1 - 2 | 453 | 148 | |
| AY079389.1 | - | 1 - 2 | 299 | 148 | ||
Set Membership: |
|